Eppendorf® Conical Tubes with SnapTec® Cap
Supplier: EPPENDORF
0030118.405 0030118.502 0030118.413 0030118.421
EPPE0030118.405EA 0 SGD
EPPE0030118.405 EPPE0030118.502 EPPE0030118.413 EPPE0030118.421
Eppendorf® Conical Tubes with SnapTec® Cap
Tubes Centrifuge Tubes
Eppendorf® Conical Tubes with SnapTec® caps offer secure sample containment for diverse applications. The innovative SnapTec® Cap ensures easy, secure sealing, making them ideal for molecular biology, genomics, and more. The conical design aids optimal sample recovery, enhancing efficiency and reliability in the lab.
  • SnapTec® caps allow effortless, secure sealing for sample integrity in molecular biology and genomics
  • Conical design enhances recovery, crucial for applications like PCR, DNA/RNA isolation, and centrifugation
  • Well-suited for various lab tasks,
More Product Information