Centrifuge tubes, Eppendorf® Tubes 5,0 ml
Supplier: EPPENDORF
0030119.509
EPPE0030119.509EA 93 SGD
EPPE0030119.509
Centrifuge tubes, Eppendorf® Tubes 5,0 ml
Tubes Centrifuge Tubes
High quality polypropylene, transparent.
  • Easier processing of samples up to 5,0 ml
  • Higher yields in DNA isolation, especially with samples with limited quantity of source material
  • Convenient and safe cell culture applications
  • Easier sample access lowers the risk of contamination
  • Preparation of mastermixes and buffers
  • Less storage space needed
  • Safe cell and tissue lysis
Eppendorf tubes are the benchmark for simple and safe sample preparation from 0,5 to 2,0 ml. For larger volumes, 15 and 50 ml conical tubes are available. With medium sample volumes youMore Product Information