Conical Tubes with Screw Caps
Supplier: EPPENDORF
0030122.178 0030122.259 0030122.267 0030122.160 0030122.186 0030122.437 0030122.500 0030122.410 0030122.429 0030122.534 0030122.542
EPPE0030122.178EA 0 SGD
EPPE0030122.178 EPPE0030122.259 EPPE0030122.267 EPPE0030122.160 EPPE0030122.186 EPPE0030122.437 EPPE0030122.500 EPPE0030122.410 EPPE0030122.429 EPPE0030122.534 EPPE0030122.542
Conical Tubes with Screw Caps
Tubes Centrifuge Tubes
Eppendorf® conical tubes with screw caps offer reliable sample containment and leak-free storage. SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
  • Polypropylene (PP) tubes, High-Density Polyethylene (HDPE) screw caps
  • Ensures leak-free storage and protects sample integrity
  • Suited for a wide range of laboratory tasks, enhancing flexibility
  • High-quality materials for long-lasting, robust performance
  • Clear, visible
More Product Information