PCR microtubes
Supplier: EPPENDORF
0030124.537
EPPE0030124.537EA 0 SGD
EPPE0030124.537
PCR microtubes
Tubes PCR Tubes
PP.
  • Etched lid for labelling
  • Tight sealing
  • Easy to open
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
These thin-walled, 0,5 ml PCR tubes are designed for use with all thermal cyclers with 0,5 ml block format.
Colour: Translucent