PCR Films and Foils
Supplier: EPPENDORF
0030127.838 0030127.854 0030127.781 0030127.790 0030132.947
EPPE0030127.838EA 0 SGD
EPPE0030127.838 EPPE0030127.854 EPPE0030127.781 EPPE0030127.790 EPPE0030132.947
PCR Films and Foils
Seals Microplate Seals
Ensure airtight sealing of your PCR plates with Eppendorf® PCR films and foils. Tailor your choice to your assay needs, like the Masterclear® real-time PCR film, optimal for qPCR with its superior optical characteristics, or heat-sealing options for the best evaportion protection.
  • PCR plate seals offer excellent chemical resistance and have been tested for resistance to a variety of common laboratory substances
  • Specialised seals with optimal optical properties are available to help minimise signal loss in qPCR
Eppendorf PCR films and foils provideMore Product Information