Eppendorf® Microplates
Supplier: EPPENDORF
0030601.106 0030601.203 0030601.300 0030602.102 0030602.200 0030602.307 0030606.302 0030621.107 0030621.301 0030622.103 0030622.308
EPPE0030601.106EA 524 SGD
EPPE0030601.106 EPPE0030601.203 EPPE0030601.300 EPPE0030602.102 EPPE0030602.200 EPPE0030602.307 EPPE0030606.302 EPPE0030621.107 EPPE0030621.301 EPPE0030622.103 EPPE0030622.308
Eppendorf® Microplates
Microplates HTS Microplates
Eppendorf microplates provide exceptional stability during centrifugation with g-Safe® technology and efficient sample identification with the OptiTrack® Matrix. The design features, including RecoverMax® well design and raised well rims, ensure reliable sealing and facilitate maximum sample recovery.
  • g-Safe®: exceptional centrifugation stability up to 6000×g
  • OptiTrack® Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labelling
  • RecoverMax® well design: Optimised well geometry for minimal remaining
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001