Polypropylene microplates, 96- and 384-well
Supplier: EPPENDORF
0030627.300
EPPE0030627.300EA 5270 SGD
EPPE0030627.300
Polypropylene microplates, 96- and 384-well
Microplates HTS Microplates
These polypropylene microplates feature high transparency and easy to read alphanumeric labelling, making the identification of the correct plate or an individual well quick, easy, and less error-prone. Black and white versions are available for fluorescence or luminescence detection, offering excellent signal-to-noise ratios for improved sensitivity.
  • Highest transparency for better sample visibility
  • OptiTrack® matrix for easy and fast well identification
  • g-safe® for time saving centrifugation up to 6000×g
  • Well designed for maximum sample recovery with
More Product Information