Eppendorf® Pipette Holder System
Supplier: EPPENDORF
3116000.015 3116000.023 3116000.031 3116000.040 3116000.058
EPPE3116000.015EA 450 SGD
EPPE3116000.015 EPPE3116000.023 EPPE3116000.031 EPPE3116000.040 EPPE3116000.058
Eppendorf® Pipette Holder System
Pipette Stands
Save valuable bench space with the Eppendorf Pipette Holder System! With space for up to six single- or multi-channel pipettes, you can easily store all of your Eppendorf pipettes and multi-dispensers within easy reach. Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments as well as most of their predecessors.
  • The Eppendorf pipette holder system stores all current mechanical and electronic Eppendorf pipettes (including the Repeater®) and most of their predecessors
  • Pipette Carousel 2: Rotatable
More Product Information