Automated pipetting system, epMotion® M5073c
Supplier: EPPENDORF
5073000.400
EPPE5073000.400EA 0 SGD
EPPE5073000.400
Automated pipetting system, epMotion® M5073c
Automated Liquid Handling Systems
The M5073 automated pipetting systems is tailored for nucleic acid purification (NAP), but is flexible for other liquid handling tasks. The integrated magnetic finger and ThermoMixer module is ideal for our MagSep reagent kits, saving hands-on time and improving the reproducibility of results. The M5073 is controlled by EasyCon tablet and can be configured with CleanCap.
  • 6 ANSI/SLAS microplate positions
  • Compact control tablet with touchscreen or mouse operation, USB data transfer
  • Software assistants for simple, step-by-step application programming
  • Optical
More Product Information