MixMate® Mixer
Supplier: EPPENDORF
5353000.537
EPPE5353000.537EA 0 SGD
EPPE5353000.537
MixMate® Mixer
Shakers Microplate Shakers
Mix samples completely and reliably with the Eppendorf® MixMate. 96 and 384-well plates and reaction tubes are mixed by the 2DMix-Control technology even at the microliter scale. The additional vortex function has a 2 s run timer allowing processing your next sample without restarting.
  • High-speed mixer (3000 rpm) for fast and efficient mixing of small volumes in all common vessel formats up to 50 ml
  • 2DMix-Control technology (planar mixing orbit without vertical movement) for controlled mixing and reproducible results in seconds
  • Anti-spill technology for
More Product Information