SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
5366000.021 5307000.000 5309000.007 5360000.038 5361000.031 5362000.035 5363000.039 5306000.006 5365000.028 5364000.024 5310000.002 5316000.004
EPPE5366000.021EA 0 SGD
EPPE5366000.021 EPPE5307000.000 EPPE5309000.007 EPPE5360000.038 EPPE5361000.031 EPPE5362000.035 EPPE5363000.039 EPPE5306000.006 EPPE5365000.028 EPPE5364000.024 EPPE5310000.002 EPPE5316000.004
SmartBlocks for ThermoMixer® C and ThermoStat C
Block Heaters Blocks
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
  • Optimised block design supports optimal temperature transfer to sample to achieve excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Insulated thermoblocks for
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001