ThermoMixer® C Thermomixer
Supplier: EPPENDORF
5382000.015 5382000.031
EPPE5382000.015EA 0 SGD
EPPE5382000.015 EPPE5382000.031
ThermoMixer® C Thermomixer
Shakers Shaking Incubators
The all-in-one for heating/cooling and mixing samples and reactions. The Eppendorf® ThermoMixer C convinces users with its quick and easy-to-exchange SmartBlocks for all common laboratory vessels. There is almost no limit to the variety of applications waiting for you.
  • Mixing, heating, and cooling in one instrument for high flexibility
  • All common vessels and plate formats from 5 µl to 50 ml by different Eppendorf SmartBlocks for standard applications
  • Ideal for cell thawing when using Eppendorf SmartBlock cryo thaw
  • Anti-spill technology prevents lid wetting
More Product Information