ThermoStat C
Supplier: EPPENDORF
5383000.019 5383000.035
EPPE5383000.019EA 0 SGD
EPPE5383000.019 EPPE5383000.035
ThermoStat C
Block Heaters Block Heater/Coolers
The Eppendorf® ThermoStat C allows you to heat or cool almost all common vessel and plate formats. Fast heating and cooling saves time. Condensation of small volumes can be prevented by using the Eppendorf® ThermoTop (optional), a heated lid with condensation-protect technology.
  • Heating and active cooling in all common vessel and plate formats from 5 µl to 50 ml for high flexibility
  • Broad range of Eppendorf SmartBlocks enables high vessel flexibility
  • Excellent temperature accuracy for safe sample handling
  • Very fast cooling rates of up to 5 °C/min, reducing
More Product Information