Microcentrifuge, refrigerated, 5427 R (General Lab Product)
Supplier: EPPENDORF
5409000.260
EPPE5409000.260EA 17520 SGD
EPPE5409000.260
Microcentrifuge, refrigerated, 5427 R (General Lab Product)
Centrifuges Microcentrifuges
This fast running microcentrifuge is designed for maximum application flexibility and offers a versatile choice of rotors in order to serve different user demands.
  • Short-Spin function allows for quick short centrifugation with pre-selectable rotation speed
  • Very quiet operation due to further improved sound insulation
  • FastTemp for rapid pre-cooling of the centrifuge, e.g. from 23 to 4 °C in approx. 11 minutes
  • Continuous cooling keeps the temperature after the run constant
  • Alternatively to continuous cooling: ECO-shut off of the compressor (adjustable after 1
More Product Information