Rotors for Centrifuges, 5910 Ri
Supplier: EPPENDORF
5895155.006 5895160.000 5895170.005 5895171.001 5895180.000 5895181.007
EPPE5895155.006EA 0 SGD
EPPE5895155.006 EPPE5895160.000 EPPE5895170.005 EPPE5895171.001 EPPE5895180.000 EPPE5895181.007
Rotors for Centrifuges, 5910 Ri
Centrifuges Rotors
Eppendorf Rotors for Centrifuge 5910 Ri - precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001