Biospectrophotometers, BioSpectrometer® basic
Supplier: EPPENDORF
6135000.009 6135000.025 6135000.904
EPPE6135000.009EA 11747.08 SGD
EPPE6135000.009 EPPE6135000.025 EPPE6135000.904
Biospectrophotometers, BioSpectrometer® basic
Spectrophotometers and Photometers Biophotometers
The BioSpectrometer basic are designed for measurements in the UV and visible range.
  • Xenon flash lamp with very long service life
  • Guided software process to minimise errors
  • Automatic evaluation and storage of measurement data with clear presentation of results
  • Integrated data processing option for specific methods
  • Two-wavelength method with substraction or division evaluation
  • Integrated application and results memory, self-test and calibration history
Spectra can be recorded and wavelength measurements can be made.
Applications include nucleic acidMore Product Information