Fluorescence spectrophotometer, BioSpectrometer®
Supplier: EPPENDORF
6137000.006 6137000.022
EPPE6137000.006EA 0 SGD
EPPE6137000.006 EPPE6137000.022
Fluorescence spectrophotometer, BioSpectrometer®
Spectrophotometers and Photometers Fluorescence Spectrophotometers
The fluorescence BioSpectrometer® offers the functions of a UV/Vis spectrophotometer plus the option to determine very low concentrations of biomolecules using fluorescent dyes.
  • Combination of absorption and fluorescence measurements
  • Sensitive nucleic acid and protein quantification via fluorescence intensity
  • Absorbance measurements for one or more wavelengths, recording of wavelength scans
  • Pre-programmed freely selectable absorption and fluorescence applications
The fluorescence unit increases the measuring range by 1000 for detecting DNA.