Cleanroom anti-fatigue mats, Infinity Bubbles
Supplier: ERGOMAT
IN0304-W IN0420-W IN0420-B IN0412-W IN0412-B IN0406-W IN0406-B IN0320-W IN0320-B IN0306-B IN0304-B IN0220-W IN0220-B IN0209-W IN0209-B IN0206-W IN0206-B IN0203-B IN0306-W
ERGOIN0304-WEA 765 SGD
ERGOIN0304-W ERGOIN0420-W ERGOIN0420-B ERGOIN0412-W ERGOIN0412-B ERGOIN0406-W ERGOIN0406-B ERGOIN0320-W ERGOIN0320-B ERGOIN0306-B ERGOIN0304-B ERGOIN0220-W ERGOIN0220-B ERGOIN0209-W ERGOIN0209-B ERGOIN0206-W ERGOIN0206-B ERGOIN0203-B ERGOIN0306-W
Cleanroom anti-fatigue mats, Infinity Bubbles
Floor Mats Traction and Anti-Fatigue Floor Mats
The Infinity Bubble mat has the comfort of a polyurethane mat and easy clean properties due to its closed-cell and non-porous top surface. It is UV and chemical resistant (except for organic solvents).
  • Upper side with bubbles
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Virgin raw material guarantee durability and ergonomic benefits
  • Chemical and UV exposure will not affect the state of the top of the mat
  • Closed cell and non-porous surface allows easy wash down and cleaning
  • Solid mat throughout, no hollow bubbles that collapse
  • Anti-static,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable