Folding precision magnifiers
Supplier: ESCHENBACH OPTIK
117612
ESCH117612EA 173 SGD
ESCH117612
Folding precision magnifiers
Magnifiers
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
  • Protective case can serve as a handle
  • Provide distortion-free images
  • Chrome plated metal
Order Now
SPECIFICATIONS
Environmentally Preferable