Kit protein dotMETRIC with spot application device (> 300 assays). Protein assay for rapid estimation of protein concentration with only a single micro liter of protein sample. Test strip is spotted and results are visible from 2-8 minutes with no pr 1 * 1 KIT
Supplier: G-Biosciences
Kit protein dotMETRIC with spot application device (> 300 assays). Protein assay for rapid estimation of protein concentration with only a single micro liter of protein sample. Test strip is spotted and results are visible from 2-8 minutes with no pr 1 * 1 KIT
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.