Skimmed milk butyrometer, without stopper
Supplier: GERBER FUNKE
3160
GERB3160EA 70.5 SGD
GERB3160
Skimmed milk butyrometer, without stopper
Butyrometers
According to Sichler`s method, with rounded scale.