Shaking water baths
Supplier: GERBER FUNKE
3550
GERB3550EA 0 SGD
GERB3550
Shaking water baths
Baths Shaking Water Baths
Shaking water bath, stainless steel. PID controller with PT-100 temperature sensor.
Delivery information: Supplied with temperature stand and 18 tubes.