Rack for solubility index tube
Supplier: GERBER FUNKE
3636
GERB3636EA 117 SGD
GERB3636
Rack for solubility index tube
Racks Tube Racks
PP, white.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean