Water bath, digital, WB-436 D
Supplier: GERBER FUNKE
3707
GERB3707EA 3320 SGD
GERB3707
Water bath, digital, WB-436 D
Baths Water Baths, Unstirred
Digital water bath with stainless steel inner and outer casing. Heating elements are located separately within the casing. Ideal for dairy use, wide range of accessories is available, it is recommended to use distilled water with the bath.
  • Digital temperture control and selectable display of set and actual temperature
  • PT100 sensor
  • Timer 1 to 99 min with audible alarm
  • Overheating protection (even when bath is empty)