Universal water bath, analogue, WB 436-A
Supplier: GERBER FUNKE
3708
GERB3708EA 2770 SGD
GERB3708
Universal water bath, analogue, WB 436-A
Baths Water Baths, Unstirred
Analogue water bath with stainless steel inner and outer casing. Heating elements are located separately within the casing. Ideal for dairy use, wide range of accessories available. It is recommended to use distilled water with the bath.
  • Analogue temperature control with adjusting knob
  • PT100 sensor
  • Thermostatic heat controller
  • Overheating protection (even when bath is empty)
Delivery information: Includes thermometer with temperature display.