Digestion tubes for Kjeldahl
Supplier: GERHARDT
12-0301
GERH12-0301EA 2270 SGD
GERH12-0301
Digestion tubes for Kjeldahl
Tubes Digestion Tubes
Made of borosilicate glass.
  • Very good temperature resistance
  • Very good chemical resistance
Order Now
SPECIFICATIONS
Environmentally Preferable