Transport Tubes, Sterile, 10 ml
Supplier: GLOBE SCIENTIFIC
6102S-R
GLBS6102S-REA 531 SGD
GLBS6102S-R
Transport Tubes, Sterile, 10 ml
Tubes Test Tubes
Sterile 16×101 mm polypropylene (PP) transport tube with separate high-density polyethylene (HDPE) red screw cap. These leak-resistant tubes are tested to 70 Kpa and gamma sterilised. The self-standing tubes feature rounded bottoms with skirt.
  • Working volune: 10 to 13 ml
  • Brim capacity: 15 ml
  • Sterilised by irradiation
  • Self-standing
  • Non-graduated
The tubes are produced from polypropylene and are RNase, DNase and pyrogen free.
The included screw caps feature an interior recess that will clasp the back end of a swab for safety and convenience whenMore Product Information