Stirling Ultracold ULT25NEU Portable/Deployable Ultra-Low Temperature Freezer
Supplier: Stirling Ultracold
ULT25NEU-PWR
GLCLULT25NEU-PWREA 23090 SGD
GLCLULT25NEU-PWR
Stirling Ultracold ULT25NEU Portable/Deployable Ultra-Low Temperature Freezer
Freezers Ultra Low Temperature Freezers
The ULT25NEU is the only portable ULT freezer solution available today for bringing ULT storage to the patient. This small lightweight, and efficient portable −80 °C freezer is ideal for clinical trials and biologic drug delivery.
  • Portable, reliable, continuous ULT operation
  • Weighs only 21 kg, can easily and inexpensively be shipped anywhere in the world as a parcel
  • Available with optional SenseAnywhere wireless temperature monitoring
  • 100% natural refrigerants
  • No compressors to fail, only two moving parts
  • Built-in universal power supply, so can plug into
More Product Information