Digital block heater, BTD
Supplier: GRANT INSTRUMENTS
BTD
GRANBTDEA 3340 SGD
GRANBTD
Digital block heater, BTD
Block Heaters Digital Block Heaters
Compact digital block heater with fixed circular block. Unit features digital control, two line display allowing simple and accurate setting of temperature and time, plus display of current status while in operation. The aluminium block is housed in durable and easy to clean, chemically resistant plastic. Unit has a 0,1 °C display resolution.
  • Operating temperature range is ambient +5 to 100 °C
  • Block accommodates up to 49 samples simultaneously: 24×1,5/2,0 ml, 15×0,5 ml and 10×0,2 ml microtubes
  • Timer with audible alarm, for operation from 1 min to 96 h
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable