Refrigerated coolers, CGR series
Supplier: GRANT INSTRUMENTS
C1GR
GRANC1GREA 8770 SGD
GRANC1GR
Refrigerated coolers, CGR series
Coolers Benchtop Coolers
Refrigerated coolers are primarily used for counter-cooling when used with a heated circulating bath or for rapidly cooling fluids down to low temperatures.
  • Coil fits beneath bridge plate of baths without reducing working space
  • Fast cooling rate: Alternative to cooling with dry ice or tap water
  • Nickel-plated copper coil with a 925 mm flexible pipe connection
The C1GR coil is suitable for use with all sizes of bath/circulators, whilst the larger height of the C2GR coil makes it suited for larger baths only.
The cooling coil may be continuouslyMore Product Information