Vacuum aspiration systems with trap flask
Supplier: GRANT INSTRUMENTS
FTA-1
GRANFTA-1EA 1850 SGD
GRANFTA-1
Vacuum aspiration systems with trap flask
Pumps Vacuum Aspiration Systems
All in one system with integrated pump designed for routine aspiration of the supernatent alcohol/buffer from the walls of microtubes during DNA/RNA purification and other macromolecule reprecipitation techniques. An ideal personal tool for independent operation away from an in-line lab vacuum supply.
  • Trap flask volume 1 L
  • Fitted with hydrophobic microbiological filter
  • Small, compact and easy to use
  • Perfect for small volume aspiration
  • The hydrophobic microbiological suction filter eliminates the risk of contamination from the trap flask
  • Built-in micro-
More Product Information