Magnetic stirrers, MMS-3000
Supplier: GRANT INSTRUMENTS
MMS-3000
GRANMMS-3000EA 985 SGD
GRANMMS-3000
Magnetic stirrers, MMS-3000
Stirrers Magnetic Stirrers
Mini magnetic stirrer with stainless steel plate. The stirrer is ideal for pH metering, extraction and dialysing with small quantities of substances.
  • Quiet operation and small size means the unit fits in easy on the labarory bench
  • Graduated dial to set stirring speed
  • Suitable for stirring samples with viscosity up to 1170 mPa
Delivery information: Supplied with cylindrical shaped magnetic stirring bar and detachable stand which enables sensors to be inserted into the sample.