Digital block heater/coolers, PCH-1, PCH-2 and PCH-3
Supplier: GRANT INSTRUMENTS
PCH-1 PCH-3
GRANPCH-1EA 4210 SGD
GRANPCH-1 GRANPCH-3
Digital block heater/coolers, PCH-1, PCH-2 and PCH-3
Block Heaters Block Heater/Coolers
The Grant bio block heater/coolers are compact, flexible and easy to use – they are ideal for the rapid heating and cooling of microtubes, and a very effective tool for DNA sample preparation for denaturing and electrophoresis. There is a choice of three models: The PCH-1 has a capacity of up to 32 microtubes in a combination of two sizes (12×1,5 ml plus 20×0,5 ml), whereas the PCH-2 can accommodate up to 20 microtubes of one size (1,5 ml), and the PCH-3 can hold up to 20×2 ml microtubes.
  • Powerful Peltier cooler for rapid heating and cooling
  • Temperature stability is ±0,1 °C
  • Two-
More Product Information