Thermoshaker with cooling, for microtubes and microplates, PCMT
Supplier: GRANT INSTRUMENTS
PCMT
GRANPCMTEA 4980 SGD
GRANPCMT
Thermoshaker with cooling, for microtubes and microplates, PCMT
Shakers Shaking Incubators
Compact thermoshaker with variable speed and temperature. This heating and cooling thermoshaker provides simultaneous or independent heating and mixing functions. The unit is ideal for DNA, RNA extractions, sample preparation, enzymatic reactions, processes and extractions of metabolites from cellular material.
  • Fast heating and cooling times 4 to 100 °C, and mixing speed of 250 to 1400 min⁻¹
  • Even mixing across the block and temperature uniformity of ±0,1 °C at 37 °C
  • Digital timer, 1 min to 96 h, with audible alarm
  • 2 line LCD display shows preset and actual
More Product Information