3 Dimensional platform rotator, PS-3D
Supplier: GRANT INSTRUMENTS
PS-3D
GRANPS-3DEA 1440 SGD
GRANPS-3D
3 Dimensional platform rotator, PS-3D
Shakers Rotating/Rocking Shakers
The compact PS-3D variable speed, fixed angle 3D rotator is ideally suited for personal use. Applications include mixing in tubes, culture flasks, gel staining, destaining, antibody staining, washes, hybridisations, and blots. The reliable motor produces regulated and reproducible rotating throughout the speed range.
  • Gentle movement keeps fragile tissue intact during incubations
  • Can be used at 4 to 40 °C (80% relative humidity, non-condensing), in an incubator or cold room
Delivery information: Supplied with non-slip mat, optional dimpled mat is available separately.