Multifunction 3 dimensional rotator, PS-M3D

Supplier: GRANT INSTRUMENTS
PS-M3D
GRANPS-M3DEA 3040 SGD
GRANPS-M3D
Multifunction 3 dimensional rotator, PS-M3D
Shakers Rotating/Rocking Shakers
The PS-M3D is a fixed angle, variable speed, multifunction 3D rotator providing variable mixing options in a compact unit. Applications include immunoprecipitation, tissue culture treatments, gel staining, destaining, antibody staining, washes, hybridisations and blots. The stepper motor produces regulated and reproducible rotating throughout the speed range. The PS-M3D provides all that is needed for mixing different sized particles.

  • Multifunction: Rotation, reciprocation and vibration
  • All actions can be set for continuous or timed operation
  • Individual
More Product Information
Order Now

SPECIFICATIONS

Model PS-M3D
Environmentally Preferable
Shaking motion 3D Orbital, reciprocal, vibration
Speed 1 - 100 min⁻¹
Angle
W×D×H 225×170×125 mm
Turning angle - reciprocal mode 1 - 360° (in steps of 30°)
Turning angle - vibration mode 1 - 5°
No. of cycles 1 - 125
Platform W×D 235×235 mm*
Weight 2 kg
Max. load 1 kg
Timer 1 - 250 s (360°/reciprocal mode)
1 - 5 s (360°/vibration mode)
Input power 4,6 W (External power supply: 12 V, 500 mA)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR