Orbital multi-platform shaker, PSU-20i
Supplier: GRANT INSTRUMENTS
PSU-20I
GRANPSU-20IEA 6930 SGD
GRANPSU-20I
Orbital multi-platform shaker, PSU-20i
Shakers Orbital Shakers
Powerful and efficient microprocessor-controlled, multifunctional orbital shaker with rotation, reciprocation and vibration. The shaker has a reliable direct drive system, accommodates loads up to 8 kg, and enables mixing optimisation whether in flasks, beakers, Petri dishes or other laboratory vessels. The PSU-20i is easy to operate, with simple set-up of multi-segment programs via push buttons and the 2-line LCD status display.
  • All mixing functions – orbital motion, reciprocation and vibration – can be set for continuous or timed operation (1 min to 96 h)
More Product Information