Vortex mixer, PV-1
Supplier: GRANT INSTRUMENTS
PV-1
GRANPV-1EA 877 SGD
GRANPV-1
Vortex mixer, PV-1
Shakers Vortex Shakers
For gentle through to vigorous mixing and pellet resuspension in 0,5, 1,5, 15 and 50 ml tubes. The PV-1 vortex mixer is so small that it will fit in your hand, with no compromise in performance.
  • Variable speed
  • Continuous or touch operation
  • Pressure-sensitive cup for tubes up to 28 mm diameter
  • Rubber suction pads prevent walking and absorb vibration
  • For use at 4 to 40 °C ambient (80% relative humidity, non-condensing) in a cold room or incubator