Accessories for digital block heaters, QBA, QBD, QBH
Supplier: GRANT INSTRUMENTS
QBEP
GRANQBEPEA 345 SGD
GRANQBEP
Accessories for digital block heaters, QBA, QBD, QBH
Block Heaters Digital Block Heaters
Tough clear polycarbonate covers maintain visibility while preventing anyone from accidentally touching a hot block.
  • Protect against splashing and are particularly recommended for use during operation at high temperatures
Order Now
SPECIFICATIONS
Environmentally Preferable