Water baths, unstirred, SUB Aqua Pro series
Supplier: GRANT INSTRUMENTS
SAP12 SAP18 SAP2 SAP26 SAP2S SAP34 SAP5
GRANSAP12EA 3440 SGD
GRANSAP12 GRANSAP18 GRANSAP2 GRANSAP26 GRANSAP2S GRANSAP34 GRANSAP5
Water baths, unstirred, SUB Aqua Pro series
Baths Water Baths, Unstirred
High quality, digital unstirred water bath range with choice of models from 2 to 34 litres and a dual bath with 5 and 12 litres. Dual baths are ideal when two different temperatures are needed and space is limited. All units have stainless steel tanks and durable painted steel cases, offer fast heat-up times, excellent temperature control, easy to use controls and a bright, clear display. All baths have heater mat technology ensuring optimum working space, without a heating element in the tank they are easier to clean. The 12 litre and larger models feature a convenient drain tap on theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable