Boiling baths, SBB Aqua Plus series
Supplier: GRANT INSTRUMENTS
SBBAQUA12PLUS SBBAQUA26PLUS SBBAQUA5PLUS
GRANSBBAQUA12PLUSEA 5810 SGD
GRANSBBAQUA12PLUS GRANSBBAQUA26PLUS GRANSBBAQUA5PLUS
Boiling baths, SBB Aqua Plus series
Baths Boiling Baths
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable