Trays for Grant SUB water baths
Supplier: GRANT INSTRUMENTS
SBT36
GRANSBT36EA 0 SGD
GRANSBT36
Trays for Grant SUB water baths
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel perforated base trays enable baths to be used as an unstirred bath.