Immersion thermostats, advanced, Optima™ TX150 and TXF200
Supplier: GRANT INSTRUMENTS
T-BTL T-BTS TXF200 TX150
GRANT-BTLEA 518 SGD
GRANT-BTL GRANT-BTS GRANTXF200 GRANTX150
Immersion thermostats, advanced, Optima™ TX150 and TXF200
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The Optima™ range of advanced thermostats can be combined with plastic baths, premium stainless steel baths or refrigeration units. The TX150 and TXF200 thermostats feature a full colour QVGA TFT screen displaying actual and set temperature and clear status icons. The QVGA TFT screen, in combination with the easy to use dial and function keys, allows rapid navigation through the intuitive menu system. Both units have RS232 and USB interfaces and can be programmed via remote PC or laptop using Grant’s versatile Labwise™ software (optional). Additional features include countdown timer,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable