Immersion thermostats, general purpose, Optima™ T100 and TC120
Supplier: GRANT INSTRUMENTS
TC120 T100
GRANTC120EA 4270 SGD
GRANTC120 GRANT100
Immersion thermostats, general purpose, Optima™ T100 and TC120
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The Optima™ range of general purpose thermostats can be combined with plastic baths, premium stainless steel baths or refrigeration units. Digital microprocessor-controlled T100 and TC120 thermostats feature an easy to use rotary dial and two function keys for quick temperature setting and menu navigation. Units feature a bright, easy to view 4 digit display, 3 programmable temperature presets, low liquid protection, fixed over-temperature cut-out, visual alarm, countdown timer and user calibration facility for optimum accuracy.
  • T100 temperature range: Ambient +5 to 100 °C. Settable
More Product Information