Thermoshaker for deep well plates, TS-DW
Supplier: GRANT INSTRUMENTS
TS-DW
GRANTS-DWEA 5300 SGD
GRANTS-DW
Thermoshaker for deep well plates, TS-DW
Block Heaters Shaking Block Heaters
The TS-DW has been designed for shaking and heating deep well plates. A multisystem principle used in the design of the thermoshaker allows it to operate as three independent devices: Incubator, plate shaker and thermoshaker. Ideal for use in life science applications, molecular biology, cell biology lab, cell lysis, DNA isolation and purification, sample preparation for PCR, pellet re-suspension, or any other method where many samples need mixing in deep well plates.
  • Profiled platform for perfect plate fit and maximum heat transfer
  • Rapid heat-up speed
More Product Information