PCR UV Cabinet - DNA/RNA, double PCR Workstation, UVT-S-AR
Supplier: GRANT INSTRUMENTS
UVT-S-ARUK
GRANUVT-S-ARUKEA 12010 SGD
GRANUVT-S-ARUK
PCR UV Cabinet - DNA/RNA, double PCR Workstation, UVT-S-AR
Fume Hoods and Enclosures PCR Workstation Enclosures
Advanced benchtop UV cabinet providing aseptic conditions for a variety of biomedical and biochemical procedures. The large capacity stainless steel UV cabinet with additional space for equipment and accessories allows more comfortable and convenient working in PCR working. With dual UV lamp protection.
  • Robust construction with large 1,2×0,52 m working area
  • UV surface irradiation – via single 25 W 254 nm open UV lamp
  • Patented built-in UV cleaner - recirculator with 25 m³/hour continuous air flow, prevents unwanted contamination and protects the user from
More Product Information