Multi-tube vortex mixer, V-32
Supplier: GRANT INSTRUMENTS
V-32
GRANV-32EA 1590 SGD
GRANV-32
Multi-tube vortex mixer, V-32
Shakers Vortex Shakers
Versatile multi-tube vortex mixer for vigorous re-suspension of cells or chemical pellets in a maximum of 32 tubes (16×1,5 ml, 8×0,5 ml and 8×0,2 ml), with the facility to mix individual tubes up to 15 ml. Typical applications include vigorous stirring and re-suspension of bacterial and yeast cells and pellets, metabolite and enzyme extraction and various DNA operations such as deproteinisation of DNA/protein complexes and purification of low molecular weight DNA/RNA fragments.
  • Easy to use - continuous or touch operation and set speed knob from 500 to 3000
More Product Information