Cryo.S™ tubes, 1 and 2 ml
Supplier: Greiner Bio-One
122278_500 122279_500 122280_500 122263_500
GREI122278_500EA 444 SGD
GREI122278_500 GREI122279_500 GREI122280_500 GREI122263_500
Cryo.S™ tubes, 1 and 2 ml
Vials
For storage in freezer or in the liquid nitrogen vapour phase. Thermal resistance of −196 to +121 °C.
  • Certified as non cytotoxic and free from endotoxins, DNase, RNase and human DNA
  • New graduation and marking field
  • Compatible with air transport
  • 1D and 2D bar codes on request
In PP, with a coloured cap, sterile (SAL 10⁻³ ).
* without writing area
** with Datamatrix code