Microplate covers, CapMats
Supplier: Greiner Bio-One
381070_50
GREI381070_50EA 0 SGD
GREI381070_50
Microplate covers, CapMats
Seals Microplate Seals
Made from ethyl vinyl acetate (EVA) or silicone, CapMats offer an alternative method for sealing 96 and 384 well microplates. All CapMats are free of RNase, DNase, and human DNA, are non-pyrogenic, and are suitable for PCR applications.
  • 96-well CapMats with round naps are suitable for sealing 1 ml and 0,5 ml 96-well MASTERBLOCK®, as well as standard 96-well PP microplates
  • 96-well CapMats with round naps can also be used to seal 96-well F-, U-, and µClear® bottom PS microplates, but not V-bottom PS microplates
  • 96-well CapMats with square naps are suitable
More Product Information