Serum collection tubes, VACUETTE®
Supplier: Greiner Bio-One
454236_1200
GREI454236_1200EA 734 SGD
GREI454236_1200
Serum collection tubes, VACUETTE®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
VACUETTE® serum tubes are coated with micronised silica particles which activate clotting when tubes are gently inverted. These tubes are used for routine clinical chemistry, immunohematology, serology and immunology testing.
These serum tubes with gel contain a barrier gel that is present in the bottom of the tube. During centrifugation, the barrier gel moves upward to the serum-clot interface where it forms a stable barrier, separating the serum from the cells.
Certifications: FDA 510 (k) cleared for therapeutic drug monitoring (TDM).
Caution: *The product(s) on this page areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable